Would you like to log in again?
VWR® Basic SMS Lab Coats
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR®
414004-357 75854-028 75854-026 414004-358 414004-367 75840-186 414004-362 414004-355 414004-360 414004-356 414004-366 414004-364 414004-365 414004-361 414004-363 414004-359
414004-359CS 227.84 USD InStock
414004-359 414004-358 414004-357 414004-367 414004-356 414004-355 414004-366 414004-365 414004-364 414004-363 75854-026 75854-028 414004-362 414004-361 414004-360 75840-186
VWR® Basic SMS Lab Coats
Disposable Garments
Lab coats are manufactured from a multilayered spunbonded-meltblown-spunbonded (SMS) fabric that offers high tensile strength while remaining soft, comfortable, and breathable.
  • Multilayer SMS material protection
  • Light fluid and particulate barrier
  • Excellent breathability
  • One breast pocket and two hip pockets
  • Serged seams
These static-dissipative garments act as a fluid and particulate barrier without sacrificing comfort or restricting movement
Garments feature a tapered collar, snap front, knit cuffs, and inset sleeves. All lab coats have one breast pocket and two hip pockets. Serged seams.
Lab coats are rigorously tested and manufactured in an ISO Certified facility under stringent process controls to ensure that each garment meets exacting quality standards and performs to specification. Lot controlled and validated through independent lab testing.
They provide light chemical splash resistance. The material is low-linting and resistant to tears and punctures. Ideally suited for use in high-class protection areas of medical, industrial, laboratory, electronic, and pharmaceutical environments.
Order Now
ORDER
SPECIFICATIONS
COMPLEMENTARY PRODUCTS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty