Would you like to log in again?
VWR® Advanced Lab Coats
Supplier: VWR
VWR®
414004-375 414004-377 414004-376 414004-380 414004-379 414004-378 89235-854
414004-379CS 234.34 USD InStock
414004-379 414004-378 414004-377 414004-376 414004-375 89235-854 414004-380
VWR® Advanced Lab Coats
Disposable Garments
Lab coats are manufactured from a specially formulated breathable microporous fabric that is exceptionally clean and durable.
  • Significant fluid and particulate barrier
  • Durable lightweight fabric
  • Excellent breathability and water vapor transmission rate
  • Soft tactile feel
These low-linting garments provide a fluid barrier and dry-particulate protection, as well as resistance to common laboratory chemicals. Appropriate for industrial, pharmaceutical, food processing, construction, and certain environmental cleanup applications.
Garments feature tapered collar, snap front, inset sleeves, elastic cuffs, and three pockets. Serged seams.
Lab coats are rigorously tested and manufactured under stringent process controls to ensure that each garment meets exacting quality standards and performs to specification. Lot controlled and validated through independent lab testing.
Certifications: Lab coats are rigorously tested and manufactured in an ISO certified facility.
Order Now
ORDER
SPECIFICATIONS
COMPLEMENTARY PRODUCTS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty