Would you like to log in again?
VWR® Basic SMS Frocks
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR®
414004-390 414004-401 414004-402 414004-391 414004-389 414004-388 414004-385 414004-386 414004-403 414004-387 414004-392
414004-403CS 243.29 USD InStock
414004-403 414004-402 414004-401 414004-389 414004-388 414004-387 414004-386 414004-385 414004-392 414004-391 414004-390
VWR® Basic SMS Frocks
Disposable Garments
Frocks are manufactured from a multilayered spunbonded-meltblown-spunbonded (SMS) fabric that offers high tensile strength while remaining soft, comfortable, and breathable.
  • Multilayer SMS material protection
  • Light fluid and particulate barrier
  • Excellent breathability
  • No pockets
  • Serged seams
These static-dissipative garments act as a fluid and particulate barrier without sacrificing comfort or restricting movement
Garments feature snap collar and snap front, raglan sleeves, and elastic cuffs. No pockets. Serged seams.
Frocks are rigorously tested and manufactured in an ISO Certified facility under stringent process controls to ensure that each garment meets exacting quality standards and performs to specification. Lot controlled and validated through independent lab testing.
They provide light chemical splash resistance. The material is low-linting and resistant to tears and punctures. Ideally suited for use in high-class protection areas of medical, industrial, laboratory, electronic, and pharmaceutical environments.
Certifications: Frocks are rigorously tested and manufactured in an ISO Certified facility under stringent process controls to ensure that each garment meets exacting quality standards and performs to specification. Lot controlled and validated through independent lab testing.
Order Now
ORDER
SPECIFICATIONS
COMPLEMENTARY PRODUCTS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty