Would you like to log in again?
VWR® Advanced Frocks
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR®
414004-408 414004-410 414004-407 414004-409 414004-406 414004-405 414004-404
414004-410CS 261.62 USD InStock
414004-410 414004-409 414004-408 414004-407 414004-406 414004-405 414004-404
VWR® Advanced Frocks
Disposable Garments
VWR® Advanced Protection Frocks are manufactured from a specially formulated breathable microporous fabric that is exceptionally clean and durable
  • Significant fluid and particulate barrier
  • Durable, lightweight fabric
  • Excellent breathability and water vapor transmission rate
  • Soft tactile feel
  • Appropriate for ISO 6 and higher
Garments feature snap collar and snap front, raglan sleeves, and elastic cuffs. Serged seams. Frocks are rigorously tested and manufactured in an ISO Certified facility under stringent process controls to ensure that each garment meets exacting quality standards and performs to specification. Lot controlled and validated through independent lab testing.
These low-linting garments provide a fluid barrier and dry-particulate protection, as well as resistance to common laboratory chemicals. Appropriate for industrial, pharmaceutical, food processing, construction, and certain environmental cleanup applications.
Order Now
ORDER
SPECIFICATIONS
COMPLEMENTARY PRODUCTS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty