Would you like to log in again?
VWR® Maximum Protection Covers
Supplier: Avantor
VWR®
414004-458 414004-459
414004-458CS 54.86 USD InStock
414004-458 414004-459
VWR® Maximum Protection Covers
Covers
Manufactured from a specially formulated breathable microporous fabric, covers are distinguished by their superior barrier protection capabilities. Provide barrier protection against fluids, biologicals, particulates, and light chemical splash.
  • Superior fluid and particulate barrier
  • Serged seams
  • Light chemical splash resistance
  • Maximum tear strength
  • Bacterial filtration efficiency 99.9%
Particle shedding Level I per Helmke Drum test.
Certifications: Covers are rigorously tested and manufactured in an ISO Certified facility under stringent process controls to ensure that each item meets exacting quality standards and performs to specification. Lot controlled and validated through independent lab testing.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty