Would you like to log in again?
VWR® Irradiated Frocks
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR®
89136-090 414004-466 89136-088 414004-464 414004-462 414004-465 414004-463
414004-466CS 284.27 USD InStock
414004-466 414004-465 414004-464 414004-463 89136-088 414004-462 89136-090
VWR® Irradiated Frocks
Disposable Garments
Irradiated frocks are manufactured from a specially formulated breathable microporous fabric that is designed to provide significant fluid and barrier protection.
  • Excellent fluid and particulate barrier
  • Extremely low level of particle shedding
  • Superior breathability and water vapor transmission rate
  • Soft, cloth-like fabric
  • Gamma irradiated to SAL of 10-4
Garments exhibit excellent water vapor transmission to optimize user comfort. Frocks are gamma irradiated to a Sterility Assurance Level (SAL) of 10-4 to address the concern of bio-burden contamination. Manufactured in an ISO Class 8 (FED-STD-209E Class 100,000/M3.5) cleanroom.
Certifications: Frocks are rigorously tested and manufactured in an ISO Certified facility under stringent process controls to ensure that each product meets exacting quality standards and performs to specification. Products are validated through independent lab testing.
Ordering information: Packed individually in sealed polyethylene bags contained within a larger carton liner. Each case is supplied with a certificate of irradiation.
Order Now
ORDER
SPECIFICATIONS
COMPLEMENTARY PRODUCTS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty