Would you like to log in again?
VWR® Maximum Protection Gowns made with Impervious Fabric
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR®
414004-469 414004-681 414004-682 414004-471 414004-470
414004-469CS 274.27 USD InStock
414004-469 414004-682 414004-681 414004-471 414004-470
VWR® Maximum Protection Gowns made with Impervious Fabric
Disposable Garments
These gowns are the cleanest and most durable within the entire VWR® Protection line of apparel.
  • Full protection from fluids and light chemical splash
  • Manufactured from fluid-impervious material
  • Lowest level of particle shedding
  • Clean, low-linting fabric
Gowns provide barrier protection against fluids, biologicals, particulates, and light chemical splash
Elastic cuffs and ties at the neck and waist ensure secure closure. Serged seams. Available in white or blue.
Gowns are rigorously tested and manufactured in an ISO Certified facility under stringent process controls to ensure that each garment meets exacting quality standards and performs to specification. Lot controlled and validated through independent lab testing.
Appropriate for industrial, pharmaceutical, food processing, and certain environmental cleanup applications.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty