Would you like to log in again?
VWR® Maximum Frocks
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR®
414004-397 414004-702 414004-698 414004-394 414004-701 414004-398 414004-700 414004-695 414004-396 414004-699 414004-696 414004-697 414004-399 414004-395
414004-700CS 297.65 USD InStock
414004-700 414004-699 414004-698 414004-697 414004-696 414004-399 414004-398 414004-695 414004-397 414004-396 414004-702 414004-701 414004-395 414004-394
VWR® Maximum Frocks
Disposable Garments
These are the cleanest and most durable frocks available within the VWR® Protection line.
  • Superior fluid and particulate barrier
  • Lowest level of particle shedding
  • Light chemical-splash resistance
  • Breathable to ensure user comfort
  • Soft, cloth-like fabric with maximum tear strength
Manufactured from a specially formulated breathable microporous fabric, these garments are distinguished by their exceptional comfort benefits and superior barrier protection capabilities
Garments feature raglan sleeves, and elastic cuffs. Available with snaps or zipper front closure. Serged seams. Color: white.
The frocks provide barrier protection against fluids, biologicals, particulates, and light chemical splash. A high vapor transmission rate keeps the wearer comfortable, even throughout extended use. Appropriate for industrial, pharmaceutical, food processing, construction, and certain environmental cleanup applications.
Certifications: Frocks are rigorously tested and manufactured in an ISO Certified facility under stringent process controls to ensure that each garment meets exacting quality standards and performs to specification. Lot controlled and validated through independent lab testing.
Order Now
ORDER
SPECIFICATIONS
COMPLEMENTARY PRODUCTS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty