Would you like to log in again?
VWR® Maximum Chemical Resistant Boot Covers
Supplier: VWR
VWR®
414005-086
414005-086CS 302.65 USD InStock
414005-086
VWR® Maximum Chemical Resistant Boot Covers
Disposable Garments
Disposable non-woven boots covers are made of 88gsm durable chemical-resistant fabric.
  • Reliable protection
  • Disposable format
  • Chemical-resistant
These garments feature ankle ties, sonic welded seams, and bonded elastic bands. Boot covers provide reliable protection in laboratory, electrical, pharmaceutical, food, industrial, and cleanroom applications. Full garment dimensions are 40.6W x 45.7L cm (16 x 18").
Certifications: ISO9001
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty