Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Eosin Y (yellowish), Signature Series™

Supplier: Epredia
Synonyms: Tetrabromofluorescein disodium saltEosin Y disodium saltEosin 2252′,4′,5′,7′-Tetrabromofluorescein disodium saltSolvent Red 43Disodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxoxanthen-9-yl)benzoateDisodium 2',4',5',7'-tetrabromo-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-bis(olate)Field's Stain B2,4,5,7-Tetrabromofluorescein disodium salt

7111 7111L 7111 7111L
41810-450CS 354.69 USD InStock
41810-450 CA89237-214 CA41810-450 89237-214
Eosin Y (yellowish), Signature Series™
Eosin Y (yellowish)
Create an ideal visual contrast to Hematoxylin with the fast-acting Epredia™ Signature Series™ Eosin-Y counterstain. Offers excellent delineation of cytoplasmic components.


Formula: C₂₀H₆Br₄Na₂O₅
MW: 691.86 g/mol
MDL Number: MFCD00005040
CAS Number: 17372-87-1
IVDLogo
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty