Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Cell Staining Buffer, Biolegend
Supplier: BioLegend
420201
420201-BLEA 118.73 USD InStock
420201-BL
Cell Staining Buffer, Biolegend
Buffers Microscopy Buffers
Cell staining buffer is an antibody diluent and cell wash buffer optimized for use in immunofluorescent staining of viable or fixed single cell suspensions.
Cell Staining Buffer contains bovine calf serum as a protein carrier to reduce non-specific binding of antibodies and fluorochrome reagents to target cells. It also contains a metabolic inhibitor, sodium azide (NaN3), to inhibit patching and capping of cell surface antigens. To prevent interference in biotin/avidin indirect staining protocols, cell staining buffer is formulated without biotin.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty