Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Intracellular Staining Permeabilization Wash Buffer (10X), Biolegend
Supplier: BioLegend
421002
421002-BLEA 186.03 USD InStock
421002-BL
Intracellular Staining Permeabilization Wash Buffer (10X), Biolegend
Buffers Microscopy Buffers
Intracellular staining permeabilization wash buffer is useful for intracellular staining procedures, e.g., in preparation of cells for staining intracellular cytokines or other proteins. Intracellular staining permeabilization wash buffer is used to permeabilize cells following fixation with intracellular staining fixation buffer.
Intracellular staining permeabilization wash buffer has been formulated to have minimal effects on cells, reduce non-specific staining and enhance the signal to noise ratio. It can be used for antibody dilutions and cell washing during intracellular staining.
The Intracellular staining perm wash buffer solution should be stored between 2 °C and 8 °C. Do not freeze.
Delivery information: It is supplied as a 10X solution and should be diluted in deionized water prior to use.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty