Would you like to log in again?
ELISA Coating Buffer (5X)
Supplier: BioLegend
421701 421701
421701-BLEA 26.39 USD InStock
421701-BL CA421701-BL
ELISA Coating Buffer (5X)
Assays ELISAs
5X concentrated carbonate buffer, pH 9.5
ELISA Coating Buffer(5X) must be diluted to 1X working solution with D.I. water prior to ELISA coating procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty