Would you like to log in again?
Plate, Round Well, J.T.Baker®
Supplier: Ritter, part of Avantor
J.T.Baker®
43001-10161 43001-00161 43001-11161 43001-00101 43001-01161
43001-1016CS 173.23 USD InStock
43001-1016 43001-0010 43001-0016 43001-1116 43001-0116
Plate, Round Well, J.T.Baker®
Microplates Deep Well Microplates
Made of virgin PP to minimise extractables.
  • Raised rim to prevent cross contamination and enable a better heat-welded seal
  • Also available rimless to allow insertion of vials, or when adhesive seals are used
  • Round bottom for optimal mixing and recovery
  • Suitable for use with automated sample handling systems
  • Can be stored at –80 °C
J.T.Baker Plate Round well plus with air escapes and raised rims for unfreezing samples.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty