Would you like to log in again?
J.T.Baker® Plate Square Well
Supplier: Ritter, part of Avantor
J.T.Baker®
43001-02001 43001-00201 43001-10621 43001-00661 43001-10661 43001-00621
43001-0200CS 99.3 USD InStock
43001-0200 43001-0066 43001-1066 43001-0020 43001-1062 43001-0062
J.T.Baker® Plate Square Well
Microplates Deep Well Microplates
These 96 deep square well plates, made from virgin PP, are designed to make the interchanging of plates simpler in automated systems. They are also ideal for housing both cell and bacterial culture.
  • Chemical resistant
  • V-bottom well shape maximises liquid removal and aids resuspension
  • No inner edges to interfere with magnetic bead collection
  • Raised well rims to improve heat sealing
  • Conical base aids sample concentration, reconstitution and centrifugation
For a maximum capacity utilization.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty