Would you like to log in again?
ELISA MAX™ Standard Set Human IFN-γ, BioLegend
Supplier: BioLegend
ELISA MAX™
430101 430101
430101-BLEA 328.56 USD InStock
430101-BL CA430101-BL
ELISA MAX™ Standard Set Human IFN-γ, BioLegend
Assays ELISAs
BioLegend’s ELISA MAX™ Standard Set contains the capture and detection antibodies, recombinant protein standard, and Avidin-HRP required for the accurate quantification of natural and recombinant human IFN-γ.
BioLegend’s ELISA MAX™ Standard Set is a sandwich Enzyme-Linked Immunosorbent Assay (ELISA). A human IFN-γ specific mouse monoclonal antibody is first coated on a 96-well plate. Standards and samples are added to the wells, and IFN-γ binds to the immobilized capture antibody. Next, a biotinylated mouse monoclonal anti-human IFN-γ detection antibody is added, producing an antibody-antigenantibody “sandwich”. Avidin-horseradish peroxidase is subsequently added, followed by TMB Substrate Solution, producing a blue color in proportion to the concentration of IFN-γ present in the sample. Finally, the Stop Solution changes the reaction color from blue to yellow, and the microwell absorbance is read at 450 nm with a microplate reader.
No cross reactivity was observed when this kit was used to analyze multiple human, mouse and rat recombinant proteins.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty