Would you like to log in again?
COMFORT BELT ADJ F/GVP PAPR
Supplier: 3M OCCUPATIONAL HEALTH
Comfort Belt^ Adjustable belt designed to evenly distribute the weight of a 3M* GVP-Series Belt-Mounted Powered Air Purifying Respirator (PAPR). Can accomodate waist sizes from 26 inches to 54 inches. The PAPR can be positioned in the back or on either side.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COMFORT BELT ADJ F/GVP PAPR 43100-080 7000126406 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty