Would you like to log in again?
3M™ EMI Copper Foil Shielding Tape 1182, 3M Company
Supplier: 3M
7100081931
43100-116EA 110.75 USD InStock
43100-116
3M™ EMI Copper Foil Shielding Tape 1182, 3M Company
Tapes
3M™ EMI shielding tapes are designed for applications requiring reliable point-to-point electrical contact, particularly EMI shielding, grounding and static charge draining.
  • A multitude of uses in electronic design and test laboratories for prototyping, design and troubleshooting
  • Eliminates the problem of rolls of tape lost between multiple users or kept loose in desk drawers
Foil shielding tape engineering kit enables engineers who need only a few inches of a particular tape for specifying, prototyping, troubleshooting, testing and repairing to avoid the problems and expense associated with meeting minimum order quantities.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty