Would you like to log in again?
3M EXHALATION VALVE 10/CS
Supplier: 3M OCCUPATIONAL HEALTH
Exhalation Valve^ This valve is used with the 3M* Half Facepiece Respirators 7500 Series, and also used with the 3M* Center Adapter 6864 on the 3M* Full Facepeice Respirators 6000 Series.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
3M EXHALATION VALVE 10/CS 43100-124 7000002159 Case of 10 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty