Would you like to log in again?
ELISA MAX™ Standard Set Mouse IL-12/IL-23 (p40)
Supplier: BioLegend
ELISA MAX™
431601 431601
431601-BLEA 283.4 USD InStock
CA431601-BL 431601-BL
ELISA MAX™ Standard Set Mouse IL-12/IL-23 (p40)
Assays ELISAs
Kit Contents
Mouse IL-12/IL-23 (p40) ELISA MAX™ Capture Antibody (200X)
Mouse IL-12/IL-23 (p40) ELISA MAX™ Detection Antibody (200X)
Mouse IL-12/IL-23 (p40) Standard
Avidin-HRP (1000X)
  • Sensitivity: 4 pg/mL
  • Standard Range: 7.8-500 pg/mL
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty