Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ELISA MAX™ Deluxe Set Human IL-2, Biolegend

Supplier: BioLegend
ELISA MAX™ Deluxe
431804 431804
431804-BLEA 410.49 USD InStock
431804-BL CA431804-BL
ELISA MAX™ Deluxe Set Human IL-2, Biolegend
Assays ELISAs
Kit Contents:
Human IL-2 ELISA MAX™ Capture Antibody (200X)
Human IL-2 ELISA MAX™ Detection Antibody (200X)
Human IL-2 Standard
Avidin-HRP (1000X)
Substrate Solution A
Substrate Solution B
Coating Buffer A (5X)
Assay Diluent A (5X)

  • Standard Range: 7.8-500 pg/mL
  • Sensitivity: 4 pg/mL

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty