Would you like to log in again?
Microfine Grinding Mill, MF 10, IKA® Works
Supplier: IKA
2836000 0002836001
43300-186EA 6144.57 USD InStock
43300-186 43300-188
Microfine Grinding Mill, MF 10, IKA® Works
Mills
This continuous microfine, stainless steel grinder features interchangeable grinding heads
Choose the cutting grinding head for crushing fibrous substances such as paper and vegetation, as well as plastics and other materials with a low volume weight. The impact grinding head crushes brittle and hard materials such as minerals and building materials.
Ordering information: A grinding head and a 304 stainless steel sieve are required for operation and must be purchased separately.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty