Would you like to log in again?
BREATHING TUBE F/RESPIRATR PK1
Supplier: 3M OCCUPATIONAL HEALTH
Breathing Tube^ This breathing tube is used on 3M* GVP Systems that have 3M* Half Facepieces 7000 Series and Full Facepieces 7800S.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
BREATHING TUBE F/RESPIRATR PK1 43300-222 7100077661 Pack of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty