Would you like to log in again?
IL-1 alpha ELISA MAX™ Standard ELISA Kit, BioLegend
Supplier: BioLegend
ELISA MAX™
433401 433401
433401-BLEA 283.4 USD InStock
CA433401-BL 433401-BL
IL-1 alpha ELISA MAX™ Standard ELISA Kit, BioLegend
Assays ELISAs
IL-1 alpha ELISA MAX™ Standard ELISA Kit reacts with Mouse
Kit Contents
Mouse IL-1α ELISA MAX™ Capture Antibody (200X)
Mouse IL-1α ELISA MAX™ Detection Antibody (200X)
Mouse IL-1α Standard
Avidin-HRP (1000X)
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty