Would you like to log in again?
IL-1 beta (Human) ELISA MAX™ Deluxe ELISA Kit, BioLegend
Supplier: BioLegend
ELISA MAX™ Deluxe
437004
437004-BLEA 443.41 USD InStock
437004-BL
IL-1 beta (Human) ELISA MAX™ Deluxe ELISA Kit, BioLegend
Assays ELISAs
Human IL-1β ELISA MAX™ Deluxe are used for the accurate recovery and measurement of analytes in complex fluids, such as serum and plasma (as well as activated cell cultures) without any interference by confounding serum factors, such as heterophilic antibody or rheumatoid factor.
  • Time-savings
  • Convenience and performance of a high-end
ELISA Max™ Set Deluxe includes pre-titrated capture antibody, pre-titrated biotinylated detection antibody, pre-titrated avidin-HRP, conveniently-sized assay calibration standards, easy-to-follow instructions, as well as, assay diluent, coating buffer, TMB substrate, and even high-affinity binding ELISA plates.
BioLegend's mouse and human IL-1β ELISA kits use antibodies raised against the mature form of IL-1β. However, because pro-IL-1β contains the mature form, these antibodies likely recognize both the pro- and the mature forms of the protein. The human IL-1β ELISA kits have been shown to equally recognize both pro- and mature forms of IL-1β.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty