Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Corning® Minimum Essential Medium (Mod.) (MEM), Corning
Supplier: Corning
Corning®
17-305-CV 50-011-PB 15-015-CV 10-009-CV 15-010-CV 51-010-PC 50-010-PC 90-009-PB 15-010-CV 10-010-CM 50-010-PC 50-011-PC 90-009-PB 10-009-CV 15-010-CM 51-010-PC 50-011-PC 15-010-CM 50-010-PB 50-010-PB 50-011-PB 17-305-CV 15-015-CV 10-010-CM 10-010-CV 10-010-CV
45000-594EA 52.14 USD InStock
45000-594 CA45000-384 CA45000-582 45000-394 45000-592 CA45000-382 CA45000-388 CA45000-586 45000-596 CA45000-584 CA45000-386 89428-906 CA11028-536 CA45000-380 CA45000-592 CA45000-394 45000-584 45000-386 45000-384 45000-582 CA45000-596 CA45000-594 45000-586 45000-388 45000-382 45000-380
Corning® Minimum Essential Medium (Mod.) (MEM), Corning
Cell Culture Media
Minimum Essential Medium (Mod.)(EMEM) based on Basal Medium Eagle (BME) formulation to aid cell propagation.
  • Low endotoxin - < 0.25EU/mL
  • Mycoplasma tested
  • Liquid media products are formulated with cell culture grade WFI water
  • Manufactured in a cGMP environment
It is a common cell culture medium developed from early work using Basal Medium Eagle (BME) with normal mammalian fibroblasts and certain subtypes of HeLa cells. When supplemented with serum, MEM has been used for cultivation of a wide variety of cells grown in monolayers, such as fibroblasts. This formulation contains Earle's salts and with or without L-glutamine.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty