Would you like to log in again?
Dulbecco's phosphate-buffered saline (DPBS), Mediatech
Supplier: Corning
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Corning®
21-031-CV 21-031-LB 20-031-CV 21-030-CM 20-030-CV 20-030-CV 21-031-LB 21-031-LX 21-031-CM 20-031-CV 21-030-CV 21-031-LX 55-031-PC 21-030-CM 55-031-PB 21-030-CV 55-031-PC 55-031-PB 21-031-CV 21-031-CM
45000-440EA 558.39 USD InStock
45000-440 45000-430 45000-444 CA45000-442 CA45000-432 45000-434 CA45000-440 45000-442 CA45000-430 45000-432 45000-426 CA45000-436 45000-438 CA45000-444 CA45000-434 45000-436 CA45000-428 45000-428 CA45000-426 CA45000-438
Dulbecco's phosphate-buffered saline (DPBS), Mediatech
Buffers Cell Culture Buffers
Dulbecco's Phosphate Buffered Saline (DPBS) is a buffer solution which can be used to maintain cell culture media in the physiological pH range of 7.0 to 7.6
  • Low endotoxin - < 0.25EU/mL
  • Mycoplasma tested
  • Liquid media products are formulated with cell culture grade WFI water
  • Manufactured in a cGMP environment
Complete Certificate of Analysis available for each production lot, along with full formulation information.
It can serve as an irrigating, transporting or diluting fluid while maintaining cell tonicity and viability for a limited period of time.
Caution: Storage temperature: Solutions 2 - 30 °C; Powder formats 2 - 8 °C
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty