Would you like to log in again?
Corning® PBS (Phosphate-Buffered Saline), Corning
Supplier: Corning
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Corning®
21-040-CV 21-040-CM 21-040-CM 21-040-CMX12 21-040-CV
71002-822PK 385.99 USD InStock
71002-822 CA45000-446 45000-448 45000-446 CA45000-448
Corning® PBS (Phosphate-Buffered Saline), Corning
Buffers Cell Culture Buffers
Phosphate buffered saline (PBS) is a buffer solution containing sodium chloride, sodium phosphate, and in some formulations potassium chloride and potassium phosphate
  • Low endotoxin - < 0.25EU/mL
  • Mycoplasma tested
  • Liquid media products are formulated with cell culture grade WFI water
  • Manufactured in a cGMP environment
PBS helps to maintain a constant pH and is isotonic and non-toxic to cells.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty