Would you like to log in again?
Corning® HBSS (Hanks Balanced Salt Solution), Corning
Supplier: Corning
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Corning®
55-022-PB 20-021-CV 21-020-CM 20-021-CV 21-023-CV 21-023-CM 21-020-CV 21-021-CV 21-021-CV 55-022-PB 21-022-CV 21-020-CM 21-023-CM 21-022-CM 20-023-CV 21-023-CV 21-021-CM 21-021-CM 20-023-CV 21-020-CV 21-022-CV 21-022-CM
45000-462PK 163.16 USD InStock
45000-462 CA45000-460 CA45000-450 45000-452 45000-460 45000-450 45000-466 CA45000-464 CA45000-454 45000-456 45000-464 CA45000-462 CA45000-452 45000-454 CA45000-458 CA45000-466 CA45000-456 45000-458 45001-152 CA45001-152 45001-101 CA10753-898
Corning® HBSS (Hanks Balanced Salt Solution), Corning
Buffers Cell Culture Buffers
Hanks' Balanced Salt Solution (HBSS) is a buffer used to maintain a physiological pH for cells maintained in non-CO₂ atmospheric conditions.
  • Low endotoxin - < 0.25EU/mL
  • Mycoplasma tested
  • Liquid media products are formulated with cell culture grade WFI water
  • Manufactured in a cGMP environment
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty