Would you like to log in again?
Corning® Yeast Extract, Powder, Corning
Supplier: Corning
Corning®
61-231-RO 61-231-RO
45000-498EA 256.25 USD InStock
CA45000-498 45000-498
Corning® Yeast Extract, Powder, Corning
Media for Microbiology Dehydrated Media for Microbiology
Yeast extract is a mixture of amino acids, peptides, water soluble vitamins and carbohydrates obtained from autolyzed yeast
This is the water soluble portion with intact B-complex vitamins and can be used as an additive for culture media.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty