Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Gentamicin sulfate solution 50 mg/ml in deionized water

Supplier: Corning

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Synonyms: NMD Inhibitor IIGaramycinGentiomycin CGentamicin sulfate saltGentamicin sulphate salt

30-005-CR 30-005-CR
45000-636PK 646.04 USD InStock
45000-636 CA45000-636
Gentamicin sulfate solution 50 mg/ml in deionized water
Gentamicin sulphate
Gentamicin works as an antimicrobial agent by binding to the 30S ribosomal subunit of certain bacteria.

It is formulated to contain 50 mg/ml gentamicin in deionized water and is recommended for use in cell culture applications at 1 ml/L.

This product is effective against mycoplasma, Gram-negative and Gram-positive bacteria and is stable at 37°C for 5 days.

Packaging: Shipping temperature: Ambient


Storage Temperature: Ambient
MDL Number: MFCD00270181
CAS Number: 1405-41-0
Order Now

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty