Would you like to log in again?
Corning® Lymphocyte Separation Medium, Corning
Supplier: Corning
Corning®
25-072-CI 25-072-CV 25-072-CV 25-072-CI
45000-728EA 195.52 USD InStock
CA45000-728 45000-728 CA45000-726 45000-726
Corning® Lymphocyte Separation Medium, Corning
Cell Separation Media
Lymphocyte Separation Medium (LSM) was originally designed for the in vitro isolation of lymphocytes from diluted whole blood
It is a sterile, iso-osmotic polysucrose and diatrizoate solution with low viscosity and a density of 1.077-1.080 g/mL at 20ºC.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty