Would you like to log in again?
Corning® SSC buffer solution 20X concentrate (saline-sodium citrate)
Supplier: Corning
Corning®
46-020-CM 46-020-CM
45001-046PK 404.91 USD InStock
45001-046 CA45001-046
Corning® SSC buffer solution 20X concentrate (saline-sodium citrate)
Buffers Electrophoresis Buffers Hybridization Buffers
20X Saline-Sodium Citrate (SSC) buffer is used in both nucleic acid hybridization and blot transfer procedures
20X is the maximum concentration used; 0.2X may be used depending on the application.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty