Would you like to log in again?
Corning® Tris, 1 M Tris-Hydrochloride Buffers, Corning
Supplier: Corning
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Corning®
46-031-CM 46-031-CM 46-030-CM 46-030-CM
45001-064PK 366.93 USD InStock
45001-064 CA45001-064 45001-066 CA45001-066
Corning® Tris, 1 M Tris-Hydrochloride Buffers, Corning
Buffers Biological Buffers
Tris-HCl buffer has many applications in molecular biology work, including use in nucleic acid purification
This product can be diluted to obtain the desired concentration.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty