Would you like to log in again?
Corning® EDTA, 0.5M pH 8.0, Corning
Supplier: Corning
Corning®
46-034-CI 46-034-CI
45001-122EA 67.06 USD InStock
45001-122 CA45001-122
Corning® EDTA, 0.5M pH 8.0, Corning
Buffers Cell Culture Buffers
EDTA (ethylenediaminetetraacetic acid) is a polyamino carboxylic acid. It is a colorless, water-soluble solid and is useful as a chelating agent due to its ability to "sequester" metal ions such as Ca2+ and Fe3+.  These cations are responsible for various undesirable effects in molecular biological and cell culture processes. After being bound by EDTA, these metal ions remain in the solution but exhibit diminished reactivity.  The use of EDTA therefore allows for greater control over many experiments.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty