Would you like to log in again?
Corning® Phosphate-Buffered Saline (PBS), 10X Concentrate, pH 7,4, Molecular Biology Grade
Supplier: Corning
Corning®
46-013-CM 46-013-CM
45001-130PK 330.65 USD InStock
45001-130 CA45001-130
Corning® Phosphate-Buffered Saline (PBS), 10X Concentrate, pH 7,4, Molecular Biology Grade
Buffers Cell Culture Buffers
Phosphate Buffered Saline (PBS) is used to maintain a physiological pH and osmolarity, two key factors in tissue culture and molecular biology research
  • Manufactured in a cGMP environment
  • Formulated with high quality WFI water
  • RNase -, DNase-, and Protease-free
  • PBS without calcium and magnesium
  • pH 7.4 ±0.1
Caution: For research use only.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty