Would you like to log in again?
Anticon® Gold HeavyWeight™ Wipes, Contec®
Supplier: Contec
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Anticon®
492224-792 492224-790 492224-789 492224-771 492224-783 492224-798
45800-060CS 972.92 USD InStock
45800-060 45800-062 45800-134 45800-058 45800-064 45800-054
Anticon® Gold HeavyWeight™ Wipes, Contec®
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Anticon® Gold HeavyWeight™ wipes are made from 100% continuous filament polyester with a double-knit, no-run interlock construction. They use patented textile technology to achieve superior sorption characteristics for critical applications.
  • Sealed edges to reduce particle and fiber generation
  • High sorptive capacity and fast adsorption rate for superior spill control
  • High tensile strength eliminates tearing and snagging in tough cleaning applications
  • Specially processed for superior cleanliness and sorbency
The proprietary sealed edges of these wipes allows the use of 100% of the wipe for cleaning without the worry of harming delicate surfaces. Anticon® Gold HeavyWeight™ wipes' low sub-micron particles and fiber counts are ideal for use in the most critical environments. Cleanroom laundered and packaged. Also available with Particle Attraction Technology (P.A.T.).
Certifications: Recommended for ISO Class 3 to 5 environments.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty