Would you like to log in again?
Anticon® SuperSorb Cleanroom Mop, Contec®
Supplier: Contec
Anticon®
492238-649
45800-080CS 1038.11 USD InStock
45800-080
Anticon® SuperSorb Cleanroom Mop, Contec®
Mops
Constructed from 100% continuous filament polyester with unique two-yarn system for superior sorbency
Designed for wet mopping in critical areas where maximum sorbency is required for applying cleaning and disinfectant solutions. Mop is durable and launderable for multiple uses. Designed for use with commercially available handles and bucket-wringer systems. Autoclavable for sterile applications.
Ordering information: Individually packed.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty