Would you like to log in again?
Count-10™ Urinalysis Standardization Systems, Myers-Stevens Group
Supplier: Myers-Stevens Group
Count-10™
10-0441 100200 10-0730
46000-002PK 151.82 USD InStock
46000-002 15173-068 46000-064
Count-10™ Urinalysis Standardization Systems, Myers-Stevens Group
Clinical Diagnostic Supplies Clinical Reagents and Kits
This system provides a complete package for the standardization of microscopic urinalysis
It utilizes calibrated labware for urine collection and sample separation, precision test slides, and versatile quality control reagents for improved test accuracy and faster throughput. From specimen collection to microscopic sediment analysis, this system eliminates the procedure-dependent variables affecting a basic screening test.
Ordering information: The Count-10 Urine Collection System comes with 500 collection cups and 500 centrifuge tubes with caps for 500 determinations. The Count-10 Urine Collection System with K-Tubes includes 500 collection cups and 500 unbreakable 12mL centrifuge tubes made with resilient K-resin for 500 determinations. The Count-10 Office Pack Urine Collection System with K-Tubes comes with 300 K-tubes, 300 Sed-Pet II™ pipets, and 300 wide-scan slides for 300 determinations.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty