Would you like to log in again?
BATTERY ACID SPILL KIT
Supplier: FYTERTECH NONWOVENS LLC
Battery Acid Spill Kit^ Contents: (2 quarts) liquid acid neutralizer, (10) 10"x12" hazmat pads, (1 pr.) nitrile gloves, (1 oz.) AQUALOCKIT* polymer, (1) poly apron, (1) trigger sprayer, (1) scoop with detachable scraper, (1) disposal bag & twist tie, (1pr.) splash goggles, red plastic carrying case.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
BATTERY ACID SPILL KIT 46000-166 440133 Case of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty