Would you like to log in again?
ES™ Series Bench Scales, Ohaus
Supplier: Ohaus
ES
71138831 71138833 71138835 71138832 71138830
46000-420EA 1151.54 USD InStock
46000-420 46000-440 46000-424 46000-432 46000-428
ES™ Series Bench Scales, Ohaus
Balances and Scales
Utility and economy come together in the ES™ Series of all-purpose bench scales
  • Weighing (4 units)
Scales are constructed of ABS plastic indicator housing and feature a stainless steel platform pan.
The low profile design of the ES™ Series scales makes them perfect for shipping, receiving, and general bench applications. Easy, two-button operation and a large display help guarantee accurate and repeatable results. The unique three-way mounting bracket allows users to mount the indicator to the scale base, above the scale platform, or on any vertical surface.
Certifications: C-UL listed.
Ordering information: Units are supplied with an AC adaptor, or may be operated on three C batteries (not included).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty