Would you like to log in again?
LITE-DRI® Absorbent, New Pig
Supplier: New Pig
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
LITE-DRI®
PLP201
46000-496PK 44.77 USD InStock
46000-496
LITE-DRI® Absorbent, New Pig
Spill Control Materials
Unlike clay, which is simply coated with liquid, fast-wicking recycled cellulose actually soaks up the spill the moment it touches
  • Absorbs and retains oils, coolants, solvents and water
  • Bag features a built-in handle for easy portability, handling and dispersal
  • Recycled cellulose is ideal when green products are desired or required
  • Can be incinerated after use to reduce waste or for fuels blending
Three times more absorbent means you'll use less material on the spill, reducing your labor and disposal costs. Silica-free composition is non-abrasive and does not pose the health hazards associated with breathing in clay dust. Non-abrasive composition won't harm finished floors or damage expensive machinery. Light-weight absorbent is easier than clay to carry and use without injury.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty