Would you like to log in again?
PIG® Skimmer Sock, New Pig
Supplier: New Pig
PIG®
104PS
46000-512PK 187.27 USD InStock
46000-512
PIG® Skimmer Sock, New Pig
Spill Control Materials
Tough, chemical-resistant polypropylene skin and filler absorb oils but repel water-based liquids.
  • Absorbs and retains oil-based liquids - including lubricants and fuels - without taking in a drop of water
Excellent for skimming oil off acid or caustic baths or for use in other acidic environments. Meets NFPA 99 standards for static decay. Attached string allows socks to be tied down, tied together or dipped into tanks. PIG Skimmer Socks float at the surface for easy retrieval.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty