Would you like to log in again?
PIG® Oil-Only Absorbent Mat Pad, New Pig
Supplier: New Pig
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
MAT426 MAT403 MAT455 MAT415 MAT460 MAT423
89073-842CS 106.63 USD InStock
89073-842 89073-854 89073-852 46000-514 89205-920 89073-846
PIG® Oil-Only Absorbent Mat Pad, New Pig
Spill Control Materials
Eight layers of 100% polypropylene are thermally bonded to give PIG Mat excellent strength; won't rip, tear or fray even when saturated.
  • Can be wrung out and incinerated after use to reduce waste or for fuels blending
  • Flame-resistant material won't burn immediately like cellulose mats; melts when exposed to high heat while on the job
  • Pads are ideal for catching drips and soaking up spills
  • Easy-tear perforations let you take only what you need, so you use less mat and save money
Heavyweight construction is highly durable and absorbent for demanding tasks and large-volume cleanups. Mediumweight construction is versatile for general, everyday tasks and cleanups. Light-weight construction is economical for small tasks and cleanups.
Exclusive dimple pattern speeds wicking of liquid throughout mat for faster, easier cleanup. Highly absorbent, fine-fiber construction won't leave behind liquids or fiber residue. Absorbs and retains oils and oil-based liquids - including lubricants and fuels - without taking in a drop of water. Bright white color makes absorbed oil easier to see; draws attention to machine leaks and clearly shows saturation level during spill response. Floats to clean up spills on water.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty