Would you like to log in again?
PIG® Original Absorbent Sock, New Pig
Supplier: New Pig
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
PIG®
PIG209 PIG218 404 204
56620-692PK 161.44 USD InStock
56620-692 46000-522 46000-520 56620-586
PIG® Original Absorbent Sock, New Pig
Spill Control Materials
Formable socks hug corners and surround machine bases to absorb machine leaks, contain puddles and help keep workers safe.
  • Absorbs and retains oils, coolants, solvents and water
  • Corncob filler is light-weight and economical to handle general, everyday tasks
  • Can be incinerated after use to reduce waste or for fuels blending
Contained corncob filler pulls liquids from the floor for maximum absorbency while making cleanup quick and easy. Polypropylene skin resists tearing; reduces dust and holds in liquid, even when saturated.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty