Would you like to log in again?
PIG® Super Absorbent Sock, New Pig
Supplier: New Pig
SUPER PIG®
PIG212 PIG211 PIG210 PIG214
56620-008CS 152.08 USD InStock
56620-008 46000-526 46000-524 56620-788
PIG® Super Absorbent Sock, New Pig
Spill Control Materials
Extra-absorbent, formable socks hug corners and walls, surround higher-volume machine leaks and contain spills to keep workers safe.
  • Absorbs and retains oils, coolants, solvents and water; not recommended for acids, bases or other corrosive liquids
  • 95% recycled cellulose filler is Excellent when green products are desired
  • Wringable for reclaiming up to 65% of absorbed liquid
  • Can be incinerated after use to reduce waste or for fuels blending
Cellulose filler is highly absorbent for large leaks and spills; super absorbency means that you replace them less often. Stitch-bonded, polypropylene skin resists bursting; reduces dust and holds in liquid, even when saturated.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty