Would you like to log in again?
PIG® HazMat Chemical Absorbent Sock, New Pig
Supplier: New Pig
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
124CR PIG301
46000-772CS 510.05 USD InStock
46000-772 56620-082
PIG® HazMat Chemical Absorbent Sock, New Pig
Spill Control Materials
Absorbs and retains most acids, bases, and unknown liquids
  • Made of 100% chemical-resistant skin and fast-wicking filler
Absorbs the widest range of acids, bases and unknown liquids, even those with high concentrations like 98% sulfuric acid and 30% sodium hydroxide. Chemical-resistant sock won't degrade or cause a dangerous reaction upon contact with corrosive spills. Excellent for quickly diking and stopping the spread of chemical spills; for diking larger chemical spills, consider our larger diameter PIG HazMat Dikes. Polypropylene skin resists chemicals and tearing; reduces dust and holds in liquid, even when saturated. Polypropylene filler is highly absorbent for containing corrosive or reactive spills.
Pink color is easily distinguishable to assure workers use the correct sock during a spill emergency. Can be incinerated after use to reduce waste.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty