Would you like to log in again?
Automated Tests, Thermo Scientific
Supplier: Thermo Scientific
1245-250 TR13421 TR12421 TR60101 TR24321 TR13421 TR60101 TR22421 TR35121 TR28321 TR28321 1265250 TR35121 TR22421
46100-248EA 226.51 USD InStock
46100-248 46100-228 CA46100-346 CA46100-228 46100-276 46100-300 CA46100-256 46100-354 CA46100-300 46100-038 46100-346 CA46100-276 46100-292 CA46100-292
Automated Tests, Thermo Scientific
Control reagents
Reagents must be stored at 2 to 8°C (36 to 46°F)
Reagents for total protein tests can be stored at 2 to 25°C (36 to 77°F).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty