Would you like to log in again?
GLOVES LGT WT SPCTRAG SM EA=PR
Supplier: HONEYWELL SAFETY PRODUCTS USA INC.
TuffShield* Gloves, Small^ Incorporates up to 25% more SpectraGuard* fiber that is low-lint, pure white, chemical-resistant and ten times stronger than steel on an equal weight basis. Excellent for knife hand applications.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
GLOVES LGT WT SPCTRAG SM EA=PR 46100-403 PF13-S Each Retrieving
Pack of 12 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty