Would you like to log in again?
GLOVES PVC 2DIP FLCE 12 PK12
Supplier: HONEYWELL SAFETY PRODUCTS USA INC.
POWERCOAT* Double-Dipped PVC Glove^ Medium. Length: 30.5cm. PVC coating repels liquid and protects hands from a wide variety of chemicals. Thermal fleece lined. Excellent cold resistance without sacrificing flexibility.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
GLOVES PVC 2DIP FLCE 12 PK12 46100-510 460-M Case of 72 Retrieving
Pack of 12 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty